Order Entry
Austria
ContactUsLinkComponent
Dry heat indicator tapes, Comply™
Dry heat indicator tapes, Comply™
Catalog # 804-0226
Supplier:  3M MEDICAL
undefined
Dry heat indicator tapes, Comply™
Catalog # 804-0226
Supplier:  3M MEDICAL

Specifications

  • Width:
    19 mm
  • Description:
    Dry heat sterilisation adhesive tape
  • Length:
    55 m
  • Seq:
    04
  • Pk:
    1 Roll

Specifications

About this item

This chemical indicator tape is designed for use as packaging tape and for the immediate identification of items exposed to dry heat processing.

A self-adhesive, semi-creped paper with an indicator ink printed as stripes across the width of the tape. The tape is white and the ink colour is pale green prior to use.

Packaging: 48 rolls per box.