Orders LinkContactUsLinkComponent
Electrode cleaning solutions
Electrode cleaning solutions
Supplier:   Hanna
Electrode cleaning solutions
Supplier:  Hanna
Certificates
Electrode cleaning solutions
3 Options Available Below
Product Details & Documents
A variety of cleaning solutions for electrodes.
  • Air tight bottle with tamper-proof seal to ensure quality
  • Expiration date and lot number clearly marked on the bottle
HI70000P is a general purpose rinsing solution that is supplied in convenient foil sachets. Electrodes and probes can become dirty from use and will produce inaccurate results. Hanna's rinsing solution eliminates impurities and residues that are left on electrodes and probes surfaces when immersed in samples during measurement or when stored incorrectly.
HI7061L is a general purpose cleaning solution that can be used for a variety of applications. Electrodes can become dirty from use and will produce inaccurate results even as they read correctly in a pH buffer. Hanna’s cleaning solutions eliminate impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly. Hanna suggests cleaning the bulb and junction of your electrode on a regular basis to ensure that the probe is always clean and to prevent any clogging of the junction.
The HI7073L is a special enzymatic cleaning solution that is specially formulated for use in applications were a pH electrode can be coated with proteins. Electrodes can become dirty from use and will produce inaccurate results even as they read correctly in a pH buffer. Hanna’s cleaning solutions eliminate impurities and residues that are left on electrode surfaces when immersed in samples during measurement and stored incorrectly. Hanna suggests cleaning the bulb and junction of your electrode on a regular basis to ensure that the probe is always clean and prevent any clogging of the junction.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 662-4195
    HI7077L Electrode Cleaning Solution for Oil and Fats, 500 ml
    HI7077L Electrode Cleaning Solution for Oil and Fats, 500 ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 51,10 €
    51,10 €
    1 * 500 mL
       51,10 €
  • 662-4110
    HI7061L General Purpose Electrode Cleaning Solution, 500 ml
    HI7061L General Purpose Electrode Cleaning Solution, 500 ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 27,70 €
    27,70 €
    1 * 500 mL
       27,70 €
  • 662-4194
    HI7073L Electrode Cleaning Solution for Proteins, 500 ml
    HI7073L Electrode Cleaning Solution for Proteins, 500 ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 51,10 €
    51,10 €
    1 * 500 mL
       51,10 €