Culture media in infusion bottles
Supplier: PHARMAMEDIA DR MULLER
Certificates
About this item
Ready to use media for sterility test. Bottles have coloured (red or green) flip-off caps.
- Identification and growth of aerobic micro-organisms
- Label contains autoclaving indicator (pink to grey-brown)
- Temperature controlled delivery
- Label with Barcode
Certifications: Medium according EP and USP
Packaging: 20 bottles on a plastic tray wrapped with shrink foil
Frequently Bought Together