ContactUsLinkComponent
Gas generators, Duran®
Gas generators, Duran®
Catalog # WITG2.989.500
Supplier:  witeg Labortechnik
undefined
Gas generators, Duran®
Catalog # WITG2.989.500
Supplier:  witeg Labortechnik
Specifications
  • Description:
    Gas generators
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Gas generators according to Kipp, Duran.
  • Tubulature at bottom
  • Jointless, total grease free apparatus
With two screw threads GL 25, one screw thread GL 45 and GL 18, with needle valve stopcock with overtwist device.
Delivery: Gas generators complete with safety funnel, caps and seals, hole disc made of sterilisable silicone, 500 ml capacity of middle bulb.