Orders LinkContactUsLinkComponent
Condenser, jacketed
Condenser, jacketed
400 mm, 29/32 NS, 29/32 NS
Catalog # QVFLGSG0107303
Supplier:   QVF LABORTECHNIK
Condenser, jacketed
Catalog # QVFLGSG0107303
Supplier:  QVF LABORTECHNIK
Condenser, jacketed
400 mm, 29/32 NS, 29/32 NS
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Cone size:
    29/32 NS
  • Jacket length:
    400 mm
  • Socket size:
    29/32 NS