Micro-volume spectrophotometer, SpectraMax® QuickDrop
735-0350
- Model:QuickDrop
- Absorbance range:−0,3..+2,5 A (–0,3...+50 A: 10 mm equivalent)
- Detector:1024 element CCD array
- Light source:Pulsed xenon lamp
- Description:Microvolume spectrophotometer
- W×D×H:290×410×190 mm
- Wavelength range:190 - 1100 nm
- Wavelength accuracy:±2 nm
- Display:177,8 mm (7") colour, capacitive touch screen
- Languages:English, Simplified Chinese, French, German, Spanish, Italian, Japanese
- Stray light:<0,5% T at 220 and 340 nm using NaNO₂
- Weight:<4 kg
- Pk:1
An affordable, one touch, full spectrum micro volume spectrophotometer, ideal for quantification of very small quantities of DNA, RNA, proteins, amino acids and peptides – a critical first step in many molecular biology, biochemistry and genetics workflows. Additionally, this unit is also suitable for many more general absorbance-based applications, including quality control and testing.
- Intuitive touch screen interface with pre-configured analysis methods simplifies data acquisition and analysis whilst still allowing the creation of customised experiments
- Fixed path length requires no calibration
- One-swipe cleaning
- Quantitate micro and large volume samples in one platform using the 0,5 µl sample or cuvette port
- Preserves precious and expensive samples and reagents
- Easily exports data to a USB drive for additional analysis
- Small footprint - no need for a dedicated PC
Short read time (<4 seconds) combined with high sensitivity and a wide spectral range provide results that are both rapid and reliable across a wide range of applications and samples.