Mounting media, Micromount®
LEIH3801731
:
- Pk:473 mL
- Description:Mounting media, Micromount®
Micromount® is a rapid-drying, xylene-based mounting medium. Its low viscosity facilitates coverslipping and prevents air bubbles forming.
- It is soluble in xylene, toluene and most xylene substitutes
An antioxidant is added to inhibit stain fading and prevent the formation of annual rings. Micromount® sets up quickly, allowing slide preparations to be handled immediately after mounting.