ContactUsLinkComponent
VWR®, Beakers, Low Form, Heavy-Duty
VWR®, Beakers, Low Form, Heavy-Duty
Catalog # 213-0481
Supplier:  VWR Collection
Supplier #:
undefined
VWR®, Beakers, Low Form, Heavy-Duty
Catalog # 213-0481
Supplier:  VWR Collection
Supplier #:
Log in for availability
Select an Option
105 mm
Select an Option
1000 ml
Select an Option
145 mm
Select an Option
07
Select an Option
6
About this item
Borosilicate 3.3 glass, with spout.
  • Good chemical and thermal shock resistance
  • Durable labelling area with matt finish and white graduated scale
  • Spout designed for optimum performance
Heavy-duty beakers have better mechanical stability than the standard products due to thicker walls. The reinforced rims increase shock resistance and reduce the risk of breakage. Due to the increased wall thickness resistance to rapid temperature changes is not as good as that of the standard product, therefore, slow, uniform heating is recommended.
Frequently Bought Together