Order Entry
Austria
ContactUsLinkComponent
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Catalog # ICNA11TWEEN201
Supplier:  MP Biomedicals
CAS Number:  
Tween® 20 (Polysorbate)
Catalog # ICNA11TWEEN201
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :1227.53 g/mol
Storage Temperature:Ambient
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986