Order Entry
Austria
ContactUsLinkComponent
Aluminium oxide acidic, MP Ecochrom™, Technical Grade A
Aluminium oxide acidic, MP Ecochrom™, Technical Grade A
Catalog # ICNA0207895
Supplier:  MP Biomedicals
CAS Number:  
Aluminium oxide acidic, MP Ecochrom™, Technical Grade A
Catalog # ICNA0207895
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    50 kg
  • Seq:
    4418

Specifications

About this item

 :
Density:3.94 g/cm³ (20 °C)
 :101.96 g/mol
Storage Temperature:Ambient
CAS Number:1344-28-1
Formula:Al₂O₃
MDL Number:MFCD00003424