Order Entry
Austria
ContactUsLinkComponent
Silicon dioxide, Ecochrom™ 32-63 µm
Silicon dioxide, Ecochrom™ 32-63 µm
Catalog # ICNA0202826
Supplier:  MP Biomedicals
CAS Number:  
Silicon dioxide, Ecochrom™ 32-63 µm
Catalog # ICNA0202826
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :60.08 g/mol
Density:2.65 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232