Orders LinkContactUsLinkComponent
Microarray scanner, GenePix® 4300A and 4400A
Microarray scanner, GenePix® 4300A and 4400A
Supplier:   Molecular Devices
Microarray scanner, GenePix® 4300A and 4400A
Supplier:  Molecular Devices
Certificates
Microarray scanner, GenePix® 4300A and 4400A
Microarray scanner, GenePix® 4300A and 4400A
2 Options Available Below
Product Details & Documents
The GenePix® 4300A and GenePix® 4400A scanners offer high resolution, high quality imaging with 3-laser excitation. Configurations include 5 μm or 2,5 μm per-pixel maximum scanning resolution, choices of up to four lasers for excitation, and sixteen emission-wavelength filters.
  • Unparalleled signal-to-noise performance
  • With integrated hardware and software
  • Non confocal optical design
Provides a powerful, flexible and easy to use solution for the acquisition and analysis of data from all types of arrays, including nucleic acids, proteins, tissues, and cells.
Product Family Options
Product Information
  • Availability
    Price
  • MLDVGENEPIX4300
    Microarray scanner, GenePix® 4300A
    Microarray scanner, GenePix® 4300A
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • MLDVGENEPIX4400
    Microarray scanner, GenePix® 4400A
    Microarray scanner, GenePix® 4400A
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST