Order Entry
Austria
ContactUsLinkComponent
96-well microplates, generic bar coded, Corning®
96-well microplates, generic bar coded, Corning®
Catalog # CORN3897BC
Supplier:  Corning
undefined
96-well microplates, generic bar coded, Corning®
Catalog # CORN3897BC
Supplier:  Corning

Specifications

  • Sterile:
    -
  • Colour:
    Black
  • Description:
    96-well microplate, V-bottom, non treated, without lid
  • Seq:
    01
  • Working volume:
    75 – 200 µl
  • Pk:
    100
  • Well volume:
    320 µl

Specifications

About this item

A line of generic bar coded 96-well plates to better meet the demands of high throughput screening.

  • Labels applied to all 4 sides of the microplate to ensure usability regardless of scanner location
  • Each microplate is specially treated to reduce the impact of static build-up
  • Code 128 bar code format ensures compatibility with most bar code scanning and software systems
  • No sequence number duplicates
  • Repeatable ANSI grading of B or better
  • Exceptional print quality and resolution