Orders LinkContactUsLinkComponent
Confidence solution, 5580
Confidence solution, 5580
Confidence solution 5580, for verifying pH, ORP, and conductivity values, 6 plastic bottles (1 pint each), 1 KIT
Catalog # YSIA005580
Supplier:   YSI ENVIRONMENTAL
Confidence solution, 5580
Catalog # YSIA005580
Supplier:  YSI ENVIRONMENTAL
CAS Number:  
Confidence solution, 5580
Confidence solution, 5580
Confidence solution 5580, for verifying pH, ORP, and conductivity values, 6 plastic bottles (1 pint each), 1 KIT
CAS Number:  
Price
246,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
This Confidence Solution® helps users to ensure probe performance of their pH, ORP, or conductivity probes.
This solution is designed for verifying measurements, not calibration. Simply immerse your multi-parameter probes in the solution and verify the pH, ORP, and conductivity values instantly. Ideal for quality checks during or after your study.
Expiration dates: Unopened=1 year, opened=3 months.
Delivery information: Case includes 6 plastic bottles (1 pint each).
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Confidence solution 5580, for verifying pH, ORP, and conductivity values, 6 plastic bottles (1 pint each)