Order Entry
Austria
Orders LinkContactUsLinkComponent
Sodium hydroxide 0.1 M (0.1 N) volumetric solution

Sodium hydroxide 0.1 M (0.1 N) volumetric solution

Catalog #
Sodium hydroxide 0.1 M (0.1 N) volumetric solution
Catalog #
Supplier:  
CAS Number:  1310-73-2
Sodium hydroxide 0.1 M (0.1 N) volumetric solution
CAS Number:  1310-73-2
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,III

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Presentation:
    Clear Colourless Liquid
  • Solubility:
    Miscible with water (D 20/4) 1,004
  • pH:
    12,7
  • Titer at 20 °C:
    0,999 - 1,001
  • Traceability :
    NIST