Pyridoxal phosphate-6-azo(benzene-2,4-disulphonic acid) tetrasodium salt hydrate ≥98% (by HPLC), solid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: PPADS tetrasodium salt hydrate, 4-[[4-Formyl-5-hydroxy-6-methyl-3-[(phosphonooxy)methyl]-2-pyridinyl]azo]-1,3-benzenedisulphonic acid tetrasodium salt
CAS Number:192575-19-2
:
Formula:C₁₄H₁₀N₃Na₄O₁₂PS₂·xH₂O
MDL Number:MFCD11046026
:599.296 g/mol
:M-Clarity™ quality level = MQ100
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:≥98% (HPLC)
- Form:Solid
- Colour:Red to Orange
- Solubility:H2O: 10 mg/ml
- Light sensitive:−20 °C
- undefined:Sensitivity
Specification Test Results
Frequently Bought Together