α-Amylase (from Aspergillus oryzae)
Supplier: Merck
Certificates
About this item
Aspergillus oryzae α-amylase enzyme catalyzes the hydrolysis of the α-1,4 glycosidic bonds in soluble starches and related subsrates. These substrates are broken down to release short oligosaccharides and α-limit dextrins. Also act as control enzyme in agar plate-based and carboxymethylcellulose-based clearing assays to screen cellulase activity.
- Used for the hydrolysis of starch to explore the the role of wheat starch in frozen dough
- Inhibit Staphylococcus aureus biofilms
- Storage temperature ranges from 2 to 8 °C
Microbial amylases are exoenzymes and are used in several industrial applications, such as production of bread, maltose syrups, and fermentation of soya sauce, miso etc. α-Amylase isolated from Aspergillus oryzae has been found to have molecular weights of 51 kDa (sedimentation and diffusion) and 49k Da (gel filtration).
Specifications
- CAS No.:9001-19-8
- Source:Aspergillus oryzae
- Form:Powder
- Enzyme name:α-Amylase
Specifications
Frequently Bought Together