Order Entry
Austria
Orders LinkContactUsLinkComponent
Isoparaffine G analytical reagent

Isoparaffine G analytical reagent

Flammable
Health
Supplier:   PanReac AppliChem
Isoparaffine G analytical reagent
Supplier:  PanReac AppliChem
Certificates
Isoparaffine G analytical reagent

2 Options Available Below

Product Details & Documents
  • CAS Number:90622-57-4
  • ADR:3,III
  •  :Filtered through a 0.2 µm filter.
  • UN:3295

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.