ContactUsLinkComponent
Cleaning cloth, low-lint, Tork Industrial
Cleaning cloth, low-lint, Tork Industrial
Catalog # SCAA190578
Supplier:  Essity
undefined
Cleaning cloth, low-lint, Tork Industrial
Catalog # SCAA190578
Supplier:  Essity
Specifications
  • For:
    W4 - Top Pak
  • Packed:
    80 wipes/pack, 5 packs/case
  • Colour:
    Blue
  • L×W:
    385×385 mm
  • Pk:
    400
Specifications
About this item
This extremely low lint cloth with anti-static properties is ideal for use with solvents when cleaning smooth surfaces. Made from 100% PP.
  • Perfect for working with sensitive surfaces due to extreme the low-lint and antistatic properties
  • Strong texture for excellent oil and water absorption
  • Tork Easy Handling™ for easier carrying, opening and disposing of packaging
  • The silicone-free cloth ensures that there are no residues on a surface
  • Food contact approved
Suitable for use in clean condition areas, printing industry, precision engineering, electronics industry.
Can be used with the Tork Folded Wiper/Cloth Dispenser that protects the refills from dirt and offers one-at-a time dispensing to reduce consumption and waste.
Dispensers
Product Information