Order Entry
Austria
ContactUsLinkComponent
Formyl peptide receptor-like 1 antagonis
Formyl peptide receptor-like 1 antagonis
  344220-5
 :  
Formyl peptide receptor-like 1 antagonis
  344220-5
 :  

 

  • Pk:
    5 mg
  • Storage conditions:
    2 - 8 °C
  • Protein synonyms:
    WRW4|H2N-WRWWWW-CONH2|FPRL1 Antagonist
  • Protein/peptide name:
    Formyl Peptide receptor-like 1 antagonis
  • Purity:
    >95%
  • Molecular formula:
    C₆₁H₆₅N₁₅O₆

 

 

The formyl peptide receptor-like 1 antagonist, WRW controls the biological activity of formyl peptide receptor-like 1 (FRPL1).

A Trp-rich hexapeptide that acts as a potent and specific antagonist of FPRL1-mediated signaling. Shown to bind to FPRL1 and interfere with the ligand-receptor interaction. Completely blocks the ligand activated [Ca²⁺]i increase, ERK activation and chemotactic migration of FPRL1-expressing RBL-2H3 cells at 10 µM. Further, inhibits Aβ₄₂ peptide-induced superoxide generation and prevents its internalization in human macrophages.