Order Entry
Austria
ContactUsLinkComponent
Probe for infrared thermometer, TFE 510
Probe for infrared thermometer, TFE 510
  620-1406
 :  EBRO
undefined
Probe for infrared thermometer, TFE 510
  620-1406
 :  EBRO

 

  • Description:
    Type T probe for TFE 510, with 600 mm cable TPE 400
  • Seq:
    01
  • Pk:
    1
  • Ø×L:
    3×120 mm