Precision thermometers, TFE 510
Supplier: EBRO
Certificates
About this item
This precision thermometer is a handy, watertight temperature measurement device for a whole range of measurement and control applications in the laboratory and in industry.
- A micro-processor controls the electronics
- Maximum measurement precision and linearity over the entire measurement range
- Power off automatically after two hours
- Measuring sensor: thermoelement
- Three times faster than comparable RTD sensor based thermometers
Thermocouple type T sensor.
Delivery: Supplied with sensor (620-1389 only), a 3 V CR Lithium battery for up to 5 years of use with charge indicator, and factory calibration certificate.
Frequently Bought Together