Orders LinkContactUsLinkComponent
Dissecting scissors
Dissecting scissors
Straight, sharp/sharp, 115 mm
Catalog # PLANT554
Marketsource
Supplier:   PLANO W. PLANNET
Dissecting scissors
Catalog # PLANT554
Supplier:  PLANO W. PLANNET
Marketsource
Dissecting scissors
Dissecting scissors
Straight, sharp/sharp, 115 mm
Price
97,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Dissecting scissors made from stainless steel.
  • With sharp tips
Specifications
  • Type:
    Straight, sharp/sharp
  • Length:
    115 mm