Rotation joint grease, ALSIROL
Catalog # 291-1320
Supplier: POLLATH
Specifications
- Description:Rotation joint grease, silicone-free, ALSIROL, 50 g
- Pk:1
Specifications
About this item
Conceived as vacuum and high vacuum grease.
- Scope of application: –40 to 200 °C
- Resistant to acids and lyes
- Vapour pressure: 0,00001 mbar
Silicon-free, no silicon derivates. No polymerisation, no sticking. Excellent adhesion and viscosity. Locked joint pairs loosened by short warming. Glass repairs are possible. Not hazardous to health in normal use. Rotation joint grease for sealing stiring systems with glass an metal stirrers. Protects rotating, precision-cut sleeves and stirring shafts from glass abrasion. Also lubricates rotary glass and metal parts.