Distillation apparatus
Distillation apparatus complete with thermometer, tripod and heating hood
NEUB9100D1160-0002
Reduced Pressure distillation apparatus according to ASTM D 1160.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
5-piece made of glass with additional 3×ball-joint clamp complete with KS 35 and NS19/38. Cylinder double wall 200 ml graduated.
- Description:Distillation apparatus complete with thermometer, tripod and heating hood