ContactUsLinkComponent
Solvent filter, bluestem glass
Solvent filter, bluestem glass
  REST26431
 :  Restek
undefined
Solvent filter, bluestem glass
  REST26431
 :  Restek
  • Description:
    Solvent filters, 15 µm frit
  • Material:
    Glass
  • Pk:
    1
This solvent filter is made up of borosilicate glass and is used to prevent the particulates and microbial growth in the solvents from entering the system.
  • Provides clean mobile phase to extend the life of columns and pump seals
  • Blue filter stem allows instant visual confirmation of upright filter orientation
  • Connects to standard Ext.Ø 3,2 mm PTFE tubing using frit adaptor
  • Borosilicate frit sits lower than conventional glass filters to draw more mobile phase from each bottle
 : Includes adaptors.
Adaptors