ContactUsLinkComponent
Ternary tee mixer
Ternary tee mixer
  REST25138
 :  Restek
undefined
Ternary tee mixer
  REST25138
 :  Restek
  • Description:
    Ternary tee mixer, stainless steel, 25 µl
  • Pk:
    1
This stainless steel ternary tee mixer is an efficient cross-flow shearing mechanism.
  • Reduced baseline noise
  • Increased sensitivity
  • Increased reaction efficiency in post-column derivatisation
  • Improved gradient accuracy for more reproducible results