ContactUsLinkComponent
Total Nitrogen test kit for AQUAfast 3700 Colorimeter and AquaMate spectrophotometers
Total Nitrogen test kit for AQUAfast 3700 Colorimeter and AquaMate spectrophotometers
Catalog # ORIOACD004
Supplier:  Thermo Orion
CAS Number:  
undefined
Total Nitrogen test kit for AQUAfast 3700 Colorimeter and AquaMate spectrophotometers
Catalog # ORIOACD004
Supplier:  Thermo Orion
CAS Number:  
Specifications
  • Chemical method:
    Digestion
  • Pk:
    50 Assays
  • Description:
    AQUAfast total nitrogen LR digestion tubes
  • Seq:
    83
  • Range:
    0,5 - 25 mg/L
  • Type:
    Digestion Tube
  • Contents:
    Set of 5: 50 TN hydroxide LR digestion tubes, 50 persulfate reagent powder packs, 50 TN reagent A powder packs, 50 TN reagent B powder packs, 50 TN Acid LR/HR reaction tubes
Specifications
About this item
Orion™ AQUAfast™ Total Nitrogen reagents in digestion tubes are designed for use with Orion™ AQUAfast™ 3700 colorimeter and Orion™ AquaMate Vis and UV-Vis spectrophotometers. Just add the sample to the digestion tube, digest sample in thermoreactor and measure.
  • Reagents are fully prepared and offer convenience and added safety for user
Digestion tubes contain the precise amount of reagent, are made of special optical glass, and fit the Orion™ AQUAfast™ COD165 thermoreactor.