Chemical resistant overalls, MICROGARD® 2300 PLUS, model 132
Supplier: Ansell
Certificates
About this item
These overalls are made from a PE barrier coating with a bi-component non woven inner layer, the combination of which provides an excellent barrier to many harmful chemicals while being lightweight and yet relatively strong and durable.
- Elasticated 3-piece hood, wrists, waist and ankles
- Two-way front zipper with resealable storm flap
- Finger loops
- Bright yellow colour for improved worker safety
- Lightweight yet relatively strong and durable
- Stitched and taped seams for increased strength and an effective barrier to liquids and particulates
- Polycoated bi-component barrier resists permeation of numerous liquid chemicals
Typical applications include: Environmental clean-up, sewage purification installations, industrial and chemical manufacturing, industrial cleaning.
Certifications: Type 3-B/4-B/5-B, EN 14126, EN 1073-2, EN 1149-5.
Specifications
- Catalog No:
- ANSEYY23T-00132-07
- ANSEYY23T-00132-06
- ANSEYY23T-00132-05
- ANSEYY23T-00132-04
- ANSEYY23T-00132-03
- ANSEYY23T-00132-02
- Model:
- 132
- 132
- 132
- 132
- 132
- 132
- Anti-static:
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial