Flame retardant and chemical resistant overalls, MICROCHEM® CFR, model 111
: Ansell
These overalls are made from a flame retardant material designed to be worn over woven thermal protective garments such as NOMEX® or PYROVATEX®, offering protection from particulates and pressurised liquid spray without compromising worker protection in the event of a flash fire.
- Two-piece hood
- Elasticated hood, wrists and ankles
- Double zip closure
- Stitched and taped seams increased strength and provide an effective liquid and particle barrier
- Flame retardant treated fabric with PVC barrier film offering protection from liquid chemicals to EN Type 3 & 4
- Highly visible bright red colour to improve worker safety
MICROCHEM® CFR offers wearers protection from liquid chemicals to EN Types 3 & 4 and peace of mind to workers in potentially explosive/flammable environments, by decreasing the risk of burn injury when worn over thermal protective workwear.
Typical applications include: Oil and petrochemicals, petroleum distribution and processing, utilities.
: Type 3/4/5, EN ISO 14116 (index 1/0/0), EN 1073-2, EN 1149-5; Cat. III
: MICROCHEM® CFR should never be worn in isolation for flame retardant protection. Always wear over the top of garments which achieve EN ISO 14116 Index 2 or above.
- Catalog No:
- ANSERD96T-00111-07
- ANSERD96T-00111-06
- ANSERD96T-00111-05
- ANSERD96T-00111-04
- ANSERD96T-00111-03
- Model:
- 111
- 111
- 111
- 111
- 111
- Anti-static:
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
Frequently Bought Together