Chemical resistant overalls, with attached gloves, Microchem® 4000, model 121
Lieferant: Ansell
Zertifikate
Über diesen Artikel
These overalls are made of multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film). They are designed to provide an exceptional barrier against organic and inorganic chemicals and biological agents.
- Double zip system helps ensure a liquid tight seal without the need for additional taping
- Elasticated 2-piece hood, waist and ankles
- Attached Ansell Barrier® gloves
- Ultrasonically welded and taped seams
- Textile-like inner improves wearer acceptance
- Permeation tested against over 180 chemicals, including chemical warfare agents
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture. Also suitable for use in EX zones.
Certifications: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5; Cat. III
Suitable for Ex-Zones
Suitable for Ex-Zones
Spezifikationen
- Catalog No:
- ANSEGR40T-00121-06
- ANSEGR40T-00121-05
- ANSEGR40T-00121-04
- ANSEGR40T-00121-03
- ANSEGR40T-00121-07
- Model:
- 121
- 121
- 121
- 121
- 121
- Anti-static:
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
Spezifikationen
Frequently Bought Together