Chemical resistant overalls, Microchem® 4000
: Ansell
These overalls are made of multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film). They are designed to provide an exceptional barrier against organic and inorganic chemicals and biological agents.
- With collar or elasticated 2-piece hood
- Double zip system helps ensure a liquid tight seal without the need for additional taping
- Double cuffs with knitted inner cuff
- Elasticated outer cuffs, waist and ankles
- Ultrasonically welded and taped seams
- Textile-like inner improves wearer acceptance
- Permeation tested against over 180 chemicals, including chemical warfare agents
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture. Also suitable for use in EX zones.
: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5; Cat. III
Suitable for Ex-Zones.
Suitable for Ex-Zones.
- Catalog No:
- ANSEGR40T-00103-05
- ANSEGR40T-00103-06
- ANSEGR40T-00111-07
- ANSEGR40T-00111-06
- ANSEGR40T-00111-05
- ANSEGR40T-00111-04
- ANSEGR40T-00111-03
- ANSEGR40T-00111-02
- ANSEGR40T-00103-04
- ANSEGR40T-00103-07
- ANSEGR40T-00103-03
- Model:
- 103, with collar
- 103, with collar
- 111, with hood
- 111, with hood
- 111, with hood
- 111, with hood
- 111, with hood
- 111, with hood
- 103, with collar
- 103, with collar
- 103, with collar
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
Frequently Bought Together