Orders LinkContactUsLinkComponent
Storage Boxes
   Sustainable Options Available
Supplier:   EPPENDORF
Storage Boxes
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
Storage Boxes
10 Options Available Below
Product Details & Documents
The storage boxes provide safe storage for your sample tubes in a space-saving design. The extensive portfolio offers a selection for tubes from 0,5 to 50 ml.
  • Made of PP for high stability in freezing applications and smooth opening and closing
  • For freezing to –86 °C
  • Autoclavable (121 °C, 20 min)
  • Alphanumeric marking of each location through laser labelling enables easy sample reference
  • Transparent lid for easy and fast sample inspection
  • Flexible and reliable labelling on the light colored box and on the 5 writing areas of the lid
  • Optimal use of freezer space due to flexible combination of the different formats
The comprehensive modular concept allows selection of the optimal box combinations for individual storage demands. The outer footprint dimensions are compatible with common freezer rack systems and the different inner grid variants provide for a perfect fit to all typical tube formats: cryogenic tubes, microcentrifuge tubes, 15 and 50 ml conical tubes and other laboratory vessels. This space-saving approach together with the application benefits resulting from the listed product features help to optimise your storage and archiving of samples.
15 and 50 ml conical tube boxes with ventilation slots for an evenly distributed freezing process. This prevents tube damage or breakage, which is frequently observed when freezing long tube formats in closed boxes.
Specifications for Product Family
Specifications
  • Catalog No:
  • 479-2167
  • 479-2168
  • 479-2171
  • 479-2172
  • 479-2169
  • 479-2170
  • 479-2175
  • 479-2173
  • 479-2174
  • EPPE0030140.729
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001.
  • Renewable Energy Certificate, ISO 14001.
  • Renewable Energy Certificate, ISO 14001.
  • Renewable Energy Certificate, ISO 14001.
  • Renewable Energy Certificate, ISO 14001.
  • Renewable Energy Certificate, ISO 14001.
  • Renewable Energy Certificate, ISO 14001.
  • Renewable Energy Certificate, ISO 14001.
  • Renewable Energy Certificate, ISO 14001.
  • Renewable Energy Certificate, ISO 14001.
Specifications
Product Family Options
Product Information
  • Compartments
    For
    W×D×H ext.
    Availability
    Price
  • 479-2167
    Hole Ø: 12,5 mm
    Hole Ø: 12,5 mm
    10×10
    100 cryotubes with internal thread
    132×132×52,8 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 31,10 €
    31,10 €
    1 * 3 ST
       31,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-2167
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact
  • 479-2168
    Hole Ø: 12,5 mm
    Hole Ø: 12,5 mm
    9×9
    Cryotubes 1-2 ml, with screw cap
    132×132×52,8 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 31,90 €
    31,90 €
    1 * 3 ST
       31,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-2168
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact
  • 479-2171
    Hole Ø: 12,5 mm
    Hole Ø: 12,5 mm
    9×9
    Cryotubes 3 ml
    132×132×76,2 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 22,10 €
    22,10 €
    1 * 2 ST
       22,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-2171
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact
  • 479-2172
    Hole Ø: 12,5 mm
    Hole Ø: 12,5 mm
    9×9
    Cryotubes 4-5 ml
    132×132×101,6 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 22,80 €
    22,80 €
    1 * 2 ST
       22,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-2172
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact
  • 479-2169
    Hole Ø: 11,9 mm
    Hole Ø: 11,9 mm
    8×8
    Tubes 1-2 ml
    132×132×52,8 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 31,90 €
    31,90 €
    1 * 3 ST
       31,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-2169
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact
  • 479-2170
    Hole Ø: 17,0 mm
    Hole Ø: 17,0 mm
    5×5
    Tubes 5 ml
    132×132×63,5 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 45,20 €
    45,20 €
    1 * 4 ST
       45,20 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-2170
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact
  • 479-2175
    Hole Ø: 17,0 mm
    Hole Ø: 17,0 mm
    5×5
    Tubes 5 ml, with screw cap
    132×132×76,2 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 22,80 €
    22,80 €
    1 * 2 ST
       22,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-2175
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact
  • 479-2173
    Hole Ø: 18,2 mm
    Hole Ø: 18,2 mm
    5×5
    Tubes 15 ml
    132×132×127 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 23,50 €
    23,50 €
    1 * 2 ST
       23,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-2173
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact
  • 479-2174
    Hole Ø: 30,3 mm (50 ml)/18,2 mm (15 ml)
    Hole Ø: 30,3 mm (50 ml)/18,2 mm (15 ml)
    3×3 (50 ml) + 2×2 (15 ml)
    Tubes 15/50 ml
    132×132×127 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 23,50 €
    23,50 €
    1 * 2 ST
       23,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-2174
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact
  • EPPE0030140.729
    Hole Ø: 30,3 mm
    Hole Ø: 30,3 mm
    3×3
    Tubes 25 ml
    132×132×88,9 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 23,80 €
    23,80 €
    1 * 2 ST
       23,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPPE0030140.729
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001.
      Low Manufacturing Impact