Order Entry
Austria
ContactUsLinkComponent
Pulse dampener, UHP
Pulse dampener, UHP
Catalog # REST26549
Supplier:  Restek
undefined
Pulse dampener, UHP
Catalog # REST26549
Supplier:  Restek

Specifications

  • Description:
    Pulse dampener, UHP
  • Pk:
    1

Specifications

About this item

The UHP pulse dampener made up of stainless steel fluid path.

  • Provides minimal flow pulsation at system pressures up to 18000 psi
  • Low dead volume reduces overall system volume

Dimensions (Ø×H): 63,5×51 mm

Wetted materials: 316 SS; PTFE; Teflon