Piston seal insertion tool, HPLC pumps
Catalog # REST21356
Supplier: Restek
Specifications
- Description:HPLC piston seal insertion tool
- Pk:1
Specifications
About this item
Piston seal insertion tool is an alternative for paper clip and screw to remove the worn seals from the HPLC pump.
- Simplify pump maintenance
- Removes old piston seal and installs new seal
- It cannot mar the surrounding metal surface of the pump housing