ContactUsLinkComponent
ESD Boot Covers, AlphaTec® 2500, model 407
Catalog # ANSEWH25B-00407-00
Supplier:  Ansell
undefined
ESD Boot Covers, AlphaTec® 2500, model 407
Catalog # ANSEWH25B-00407-00
Supplier:  Ansell
Specifications
  • Shoe size:
    42 - 46
  • Garment application:
    Cleanroom,
    Laboratory,
    Industrial
  • Colour:
    White
  • Model:
    407
  • Pk:
    150 PAIR
  • Seam type:
    Bound
  • Weight:
    65 g/m²
Specifications
About this item
Disposable overboots made from durable microporous polypropylene laminate which provides an excellent barrier to chemical spray and infective agents. With anti-slip sole made from PVC.
  • Elasticated opening
  • Adjustable shoe tie
  • Bound seams
  • ESD PVC sole
  • Unique material offering exceptional strength, liquid and particulate protection
  • Low linting reduces risk of contamination in critical areas
The fabric achieves the highest classifications for protection from biological agents in accordance with EN 14126 for penetration of blood, body fluids and blood-borne pathogens.