Water standards for Karl Fischer titration, Hydranal™
Supplier: Honeywell Chemicals
Certificates
About this item
Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application.
- Manufactured according to ISO 9001 requirements
- Report of Analysis showing the exact water content and expanded uncertainty
- Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
- Long shelf life (up to five years)
- Convenient packaging
Delivery information: Supplied with detailed instruction for use.
Caution: The recommended storage temperature for product HYDRANAL™ Water Standard is 2-8°C. The product can be transported at normal ambient temperatures. In the long term, temperatures > 20°C (if possible > 8 °C) should be avoided.
Frequently Bought Together