ContactUsLinkComponent
Apron with sleeves, MICROGARD® 2000, model 214
Apron with sleeves, MICROGARD® 2000, model 214
Catalog # ANSEWH20B-00214-07
Supplier:  Ansell
undefined
Apron with sleeves, MICROGARD® 2000, model 214
Catalog # ANSEWH20B-00214-07
Supplier:  Ansell
Specifications
  • Clothing Size:
    3XL
  • Garment application:
    Medical,
    Clinical
  • Colour:
    White
  • Seq:
    06
  • Pk:
    60
Specifications
About this item
These aprons are made from a microporous PE film with a non woven inner layer. The fabric provides low hazard liquid chemical repllence particle protection, protection from pesticides and a barrier to biological agents.
  • Can be used as a hospital gown
  • Rear Velcro fastening
  • Elasticated wrists
  • Bound seams