Raw polyurethane foam (PUF) plugs and glass holder
Supplier: Restek
Certificates
About this item
PUF plugs fits to glass holders as an alternative to pre-cleaned packed cartridges.
- Small glass holder with a stem
- Tubing connection to sampling pump
- Durable and reusable PUF glass holders
Designed for air sampling applications.
Frequently Bought Together