Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, triethylammonium acetate, 1 M solution, pH 7,0

Buffer, triethylammonium acetate, 1 M solution, pH 7,0

Buffer, triethylammonium acetate, 1 M solution, pH 7,0
Supplier:  PanReac AppliChem
Certificates
Buffer, triethylammonium acetate, 1 M solution, pH 7,0
Buffer, triethylammonium acetate, 1 M solution, pH 7,0

2 Options Available Below

Product Details & Documents

Used for the purification of oligonucleotides.

CAS No.: 5204-74-0
Molecular Formula: C₆H₁₅N · CH₃COOH
Formula Weight: 161,24 g/mol
Density: 1,001 (at 20 °C)
Physical Form: Liquid
Non-volatile Matter: Max. 0,01%
Storage Temperature: Ambient

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • A3846.0500
  • A3846.1000
  • CAS No.:
  • 5204-74-0
  • 5204-74-0

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • A3846.0500
    Triethylammonium acetate, 1M buffer solution, pH 7,0
    Triethylammonium acetate, 1M buffer solution, pH 7,0
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 117,00 €
    117,00 €
    1 * 500 mL
       117,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    A3846.0500
    CAS No.
    5204-74-0
  • A3846.1000
    Triethylammonium acetate, 1M buffer solution, pH 7,0
    Triethylammonium acetate, 1M buffer solution, pH 7,0
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 163,00 €
    163,00 €
    1 * 1 L
       163,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    A3846.1000
    CAS No.
    5204-74-0