Order Entry
Austria
Orders LinkContactUsLinkComponent
Urobilinogen 25 g/l in aqueous solution

Urobilinogen 25 g/l in aqueous solution

25 g
Catalog # A1536.0025
Supplier:   PanReac AppliChem
Urobilinogen 25 g/l in aqueous solution
Catalog # A1536.0025
Supplier:  PanReac AppliChem
CAS Number:  14684-37-8
Urobilinogen 25 g/l in aqueous solution

25 g

CAS Number:  14684-37-8
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:14684-37-8
  • Formula:C₃₃H₄₂-50N₄O₆
  • Molecular Weight:590.7 · 598.8 g/mol
Specifications

Specification Test Results

  • Form:
    Liquid (Aqueous solution)
  • Storage:
    −20 °C under argon
  • Assay:
    25 g/l
  • 4 ml contain:
    100 mg
  • pH:
    13,2±0,2 (20 °C)
  • Bilirubin:
    Max. 1%

Specifications

  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.