Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Forceps
Splinter forceps
Splinter forceps
232-0063
:
HAMMACHER KARL
Splinter forceps
232-0063
:
HAMMACHER KARL
Type:
Straight, pointed
Length:
105 mm
Seq:
03
Pk:
1
Splinter forceps with lancet tips.