ContactUsLinkComponent
Zeocin® 100 mg/ml in HEPES cell culture tested, sterile filtered
Zeocin® 100 mg/ml in HEPES cell culture tested, sterile filtered
Certificates
Zeocin® 100 mg/ml in HEPES cell culture tested, sterile filtered
Certificates
Specification Test Results
  • Quality Control:
    Each lot is thoroughly tested to ensure the absence of lot-to-lot variation.
  • Endotoxin level:
    < 1 EU/mg
  • Physicochemical characterization:
    HPLC, pH, appearance
  • Cell-culture tested:
    potency validated in Zeocin®-sensitive and Zeocin®-resistant mammalian cell lines
  • Non-cytotoxicity of trace contaminants:
    absence of long-term effects confirmed in Zeocin®-resistant cells