Real-Time PCR kit for detection of Pseudomonas aeruginosa
Lieferant: Q BIOANALYTIC
Zertifikate
Über diesen Artikel
Pseudomonas aeruginosa is tested usually with cultural methods. However, this may take several days to deliver results. Using this Real-Time PCR test you may have results already after a few hours using a DNA purification directly from filtered water without pre-enrichment. You can also use this test in combination with an appropriate pre- enrichment according to DIN EN ISO 20837. The kit is also suitable to confirm bacterial colonies on agar plates.
- TaqMan probes
- Taget in FAM channel
- Contains UNG to protect from re-amplification
- Positive DNA and negative control included
Certifications: DIN EN ISO 20838 conform detection in food and feed samples from pre-enrichment cultures or for confirmation of bacterial colonies.
Frequently Bought Together