Order Entry
Austria
ContactUsLinkComponent
Electronic rechargeable crimpers and decappers
Electronic rechargeable crimpers and decappers
  REST22360
 :  Restek
undefined
Electronic rechargeable crimpers and decappers
  REST22360
 :  Restek

 

  • Description:
    Electronic universal decapper for 11 mm crimp caps
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    1

 

 

Electronic rechargeable crimpers and decappers have a rechargeable 6,4 V battery installed. Charging the battery takes thirty minutes to two hours. The universal plug set includes U.S., European, UK, and Australian plug clips.

  • Easy to use with a comfortable grip
  • Adjustable crimping force
  • Hundreds of operations from one charge
  • Large battery allows for faster charge cycle
  • Compatible with 11 mm and 20 mm caps
 : All kits include a universal plug set.