Order Entry
Austria
Orders LinkContactUsLinkComponent
 

SuperBlock™ Blocking Buffer

SuperBlock™ Blocking Buffer
Certificates
SuperBlock™ Blocking Buffer

9 Options Available Below

Product Details & Documents

SuperBlock Blocking buffer is an optimised solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.

  • Features of SuperBlock Blocking Buffer include:
  • Blocking time - block membranes in five to ten minutes and ELISA plates in two minutes
  • Application - ELISA, western blotting, IHC biotin-free for use with streptavidin system
  • Stability - store buffer at 4 ℃ for one year store blocked plates dry for up to 12 months

Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilising them for drying and storage for later use.

 : For research use only. Not for use in diagnostic procedures.

 

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • PIER37515
  • PIER37518
  • PIER37580
  • PIER37517
  • PIER37537
  • PIER37535
  • PIER37581
  • PIER37516
  • PIER37536
  • Concentration:
  • 1X
  • 1X
  • 1X
  • 1X
  • 1X
  • 1X
  • 1X
  • 1X
  • 1X
  • For use with:
  • ELISA, Western blot
  • ELISA, Western blot
  • ELISA, Western blot
  • Western blot
  • Western blot
  • ELISA, Western blot
  • ELISA, Western blot
  • ELISA, Western blot
  • ELISA, Western blot
  • Physical form:
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Storage buffer:
  • PBS
  • PBS
  • PBS
  • PBS
  • TBS
  • TBS
  • TBS
  • PBST
  • TBST

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • PIER37515
    SuperBlock® (PBS) blocking buffer
    SuperBlock® (PBS) blocking buffer
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 340,00 €
    340,00 €
    1 * 1 L
       340,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37515
    Concentration
    1X
    For use with
    ELISA, Western blot
    Physical form
    Liquid
    Storage buffer
    PBS
  • PIER37518
    SuperBlock® (PBS) blocking buffer
    SuperBlock® (PBS) blocking buffer
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 844,00 €
    844,00 €
    1 * 5 L
       844,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37518
    Concentration
    1X
    For use with
    ELISA, Western blot
    Physical form
    Liquid
    Storage buffer
    PBS
  • PIER37580
    SuperBlock® (PBS) blocking buffer
    SuperBlock® (PBS) blocking buffer
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 64,30 €
    64,30 €
    1 * 100 mL
       64,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37580
    Concentration
    1X
    For use with
    ELISA, Western blot
    Physical form
    Liquid
    Storage buffer
    PBS
  • PIER37517
    SuperBlock® (PBS) blocking buffer- Blotting
    SuperBlock® (PBS) blocking buffer- Blotting
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 301,00 €
    301,00 €
    1 * 1 L
       301,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37517
    Concentration
    1X
    For use with
    Western blot
    Physical form
    Liquid
    Storage buffer
    PBS
  • PIER37537
    SuperBlock® (TBS blotting) blocking buffer
    SuperBlock® (TBS blotting) blocking buffer
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 317,00 €
    317,00 €
    1 * 1 L
       317,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37537
    Concentration
    1X
    For use with
    Western blot
    Physical form
    Liquid
    Storage buffer
    TBS
  • PIER37535
    SuperBlock® (TBS) blocking buffer
    SuperBlock® (TBS) blocking buffer
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 331,00 €
    331,00 €
    1 * 1 L
       331,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37535
    Concentration
    1X
    For use with
    ELISA, Western blot
    Physical form
    Liquid
    Storage buffer
    TBS
  • PIER37581
    SuperBlock® (TBS) blocking buffer
    SuperBlock® (TBS) blocking buffer
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 64,30 €
    64,30 €
    1 * 100 mL
       64,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37581
    Concentration
    1X
    For use with
    ELISA, Western blot
    Physical form
    Liquid
    Storage buffer
    TBS
  • PIER37516
    SuperBlock® T20 (PBS) blocking buffer with 0,05% Tween®20 detergent
    SuperBlock® T20 (PBS) blocking buffer with 0,05% Tween®20 detergent
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 342,00 €
    342,00 €
    1 * 1 L
       342,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37516
    Concentration
    1X
    For use with
    ELISA, Western blot
    Physical form
    Liquid
    Storage buffer
    PBST
  • PIER37536
    SuperBlock® T20 (TBS) blocking buffer with 0,05% Tween®20 detergent
    SuperBlock® T20 (TBS) blocking buffer with 0,05% Tween®20 detergent
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 342,00 €
    342,00 €
    1 * 1 L
       342,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37536
    Concentration
    1X
    For use with
    ELISA, Western blot
    Physical form
    Liquid
    Storage buffer
    TBST