Static in-line mixers, HyperShear
Supplier: Restek
Certificates
About this item
These stainless steel or PEEK in-line mixers are an efficient cross-flow shearing mechanism.
- Reduced baseline noise
- Increased sensitivity
- Increased reaction efficiency in post-column derivatisation
- Improved gradient accuracy for more reproducible results
Frequently Bought Together