Orders LinkContactUsLinkComponent
Thermometer case
Thermometer case
Wood tank thermometer cup case
Catalog # 620-0997
Supplier:   Amarell
Thermometer case
Catalog # 620-0997
Supplier:  Amarell
Thermometer case
Thermometer case
Wood tank thermometer cup case
Price
33,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
This cupcase made from wood, can be easily fixed on a wall for easier reading of the temperature.
Conforms to ASTM D1086
Zertifizierung: Conforms to ASTM D1086.
Spezifikationen
  • Description:
    Wood tank thermometer cup case