Order Entry
Austria
ContactUsLinkComponent
 
3,4-Ethylenedioxythiophene 97%
 
3,4-Ethylenedioxythiophene 97%
 

 

  • Assay (GC):
    96,0% min
  • Solubility:
    Miscible with alcohol and ether. Immsicible with water

 

Frequently Bought Together