Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt 7.444 g/l in aqueous solution, AVS TITRINORM
EDTA disodium salt 7.444 g/l in aqueous solution, AVS TITRINORM
Catalog # 88826.9011
Supplier:  VWR Chemicals
CAS Number:  
undefined
EDTA disodium salt 7.444 g/l in aqueous solution, AVS TITRINORM
Catalog # 88826.9011
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :336.21 g/mol
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672